Learn More
Chemglass Life Sciences Incubator, Digital, myTemp 65HC, Heating and Cooling, 120V
SDP
Catalog No. 5000083150
Encompass
Accurate Temp. Control from 0 to 60 ° C S. S. Internal Chamber (65L) with Light Internal Worldwide-Universal Outlet Competitively Priced with "Heat Only" Incubators Uniquely Sized for Floor-Standing Use or on Bench Tops less than 32" Tall. The myTemp 65HC Incubator is designed to provide accurate and uniform temperature control from 0 to 60 ° C. The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications. A large, digital display allows for visualization of the chamber temperature and the remaining incubation time, while an internal light is included for improved visualization of the chamber. In addition, a built in, internal outlet can be used to provide power to small instruments, such as shakers, rockers, etc. With forced air technology and stainless steel inner construction, uniform heat transfer is ensured throughout the internal chamber. When set for sub-ambient temperatures, the powerful compressor is activated, quickl
Catalog No. 50-000-83150 Supplier Chemglass Life Sciences Supplier No. CLS2457B01
Accurate Temp. Control from 0 to 60 ° C S. S. Internal Chamber (65L) with Light Internal Worldwide-Universal Outlet Competitively Priced with "Heat Only" Incubators Uniquely Sized for Floor-Standing Use or on Bench Tops less than 32" Tall. The myTemp 65HC Incubator is designed to provide accurate and uniform temperature control from 0 to 60 ° C. The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications. A large, digital display allows for visualization of the chamber temperature and the remaining incubation time, while an internal light is included for improved visualization of the chamber. In addition, a built in, internal outlet can be used to provide power to small instruments, such as shakers, rockers, etc. With forced air technology and stainless steel inner construction, uniform heat transfer is ensured throughout the internal chamber. When set for sub-ambient temperatures, the powerful compressor is activated, quickl