Learn More
Chemglass Life Sciences Incubator, 2.0 Cu. Ft., General Purpose, 120V
SDP
Supplier:  Chemglass Life Sciences CLS2440001
Encompass
Microprocessor Control / Digital LEDInterior Glass Door with Radius Corner. Unique Warm Air-Jacket Design. Heated Tempered Safety Glass Door. One Interior Outlet. Access Port. No Rivets; Seam Welded Panels and Doors. 24 Month Limited Warranty. The CLS-2440 series of general purpose incubators deliver a superior degree of temperature uniformity. This design requires no blowers or fans within the incubator chamber. Five strategically placed heating elements, including a heated outer door, ensure uniform heating while the unique air jacket design eliminates hot spots when the incubator chamber is fully loaded. The microprocessor controller achieves exceptional temperature control with no overshoot. Set point and actual chamber temperature are digitally displayed. Temperature calibration is easily performed by touch pad control. A totally independent secondary temperature controller offers the added safety of oven temperature protection.
Catalog No. 50-143-559