Learn More
Chemglass Life Sciences Incubator Genie, 120V
SDP
Catalog No. 50143356
Encompass
The Incubator Genie allows you to incubate, rotate, rock, and shake all in one compact unit. The magnetic stainless steel platform may be easily set to rock or rotate almost any container. Precise temperature control and uniformity - range from 28 degreesC to 75 degreesC which makes it ideal for hybridization. Chamber air is gently and continuously re-circulated ensuring temperature consistency and uniformity. The shatterproof window allows for inspection of samples without disturbing chamber environment. A variety of accessories are available to provide maximum application versatility. Supplied with: 1 clip plate each for 10-13mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips, non-skid tray and shelf.
Catalog No. 50-143-356 Supplier Chemglass Life Sciences Supplier No. CLS2541001
The Incubator Genie allows you to incubate, rotate, rock, and shake all in one compact unit. The magnetic stainless steel platform may be easily set to rock or rotate almost any container. Precise temperature control and uniformity - range from 28 degreesC to 75 degreesC which makes it ideal for hybridization. Chamber air is gently and continuously re-circulated ensuring temperature consistency and uniformity. The shatterproof window allows for inspection of samples without disturbing chamber environment. A variety of accessories are available to provide maximum application versatility. Supplied with: 1 clip plate each for 10-13mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips, non-skid tray and shelf.