Learn More
Research Products International Corp Incubator Genie, Benchtop
SDP
Catalog No. 501976813
Encompass
Allows to heat, rotate, rock, and shake all in one compact unit. The incubator is best for applications such as Gel and Blot staining, Cell or Bacterial Culture, as well as any application that requires mixing, washing, rocking, and temperature control of a sample. Our unique air circulation system gently and continuously guarantees temperature consistency throughout the entire chamber and mixing process.
The unit is compatible with an extensive accessory line-up.
Supplied with:
1 Clip Plate each for 10-13 mm, 15-17mm and 28-30mm Tubes
4 Bag Mounting Strips
Non-Skid Tray and Shelf
Stack-able Wire Rack
Catalog No. 50-197-6813 Supplier Research Products International Corp Supplier No. 155584
Add to Cart
Add to Cart
Request a Quote
Allows to heat, rotate, rock, and shake all in one compact unit. The incubator is best for applications such as Gel and Blot staining, Cell or Bacterial Culture, as well as any application that requires mixing, washing, rocking, and temperature control of a sample. Our unique air circulation system gently and continuously guarantees temperature consistency throughout the entire chamber and mixing process.
The unit is compatible with an extensive accessory line-up.
Supplied with:
1 Clip Plate each for 10-13 mm, 15-17mm and 28-30mm Tubes
4 Bag Mounting Strips
Non-Skid Tray and Shelf
Stack-able Wire Rack