Learn More
Darwin Chambers Company Single Door Incubator 21 cu. ft.
SDP
Supplier:  Darwin Chambers Company HT030AA
Encompass_Preferred
The HT030 is a 23.9 cubic ft single door heated incubator. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3897
Add to Cart
Request a Quote