Learn More
Darwin Chambers Company Benchtop Incubator 9 cu. ft
SDP
Catalog No. 501893896
Encompass_Preferred
The HT011 is a 11.6 cubic ft benchtop heated incubator that can be stacked w/ optional racking. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes w/ two stainless steel shelves. The chamber provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3896 Supplier Darwin Chambers Company Supplier No. HT011
Add to Cart
Add to Cart
Request a Quote
The HT011 is a 11.6 cubic ft benchtop heated incubator that can be stacked w/ optional racking. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes w/ two stainless steel shelves. The chamber provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.