Learn More
Darwin Chambers Company Triple Door Incubator 66 cu. ft.
SDP
Catalog No. 501893902
Encompass_Preferred
The HT084-SS is a 76.2 cubic ft triple door heated incubator. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3902 Supplier Darwin Chambers Company Supplier No. HT084SS
Add to Cart
Add to Cart
Request a Quote
The HT084-SS is a 76.2 cubic ft triple door heated incubator. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.