Learn More
Darwin Chambers Company Single Door Incubator 21 cu. ft.
SDP
Catalog No. 501893898
Encompass_Preferred
The HT030-SS is a 23.9 cubic ft single door heated incubator. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3898 Supplier Darwin Chambers Company Supplier No. HT030SS
Add to Cart
Add to Cart
Request a Quote
The HT030-SS is a 23.9 cubic ft single door heated incubator. Temperature range is 5°C above ambient to 50°C degrees C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.