Learn More
Darwin Chambers Company Benchtop Heat Humidity Chamber 9 cu. ft.
SDP
Catalog No. 501893881
Encompass_Preferred
The HH011 is a 11.6 cubic ft benchtop heated/humidified incubator that can be stacked w/ optional racking. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ two stainless steel shelves. The chamber provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3881 Supplier Darwin Chambers Company Supplier No. HH011
Add to Cart
Add to Cart
Request a Quote
The HH011 is a 11.6 cubic ft benchtop heated/humidified incubator that can be stacked w/ optional racking. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ two stainless steel shelves. The chamber provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.