Learn More
Darwin Chambers Company Double Door Heat Humidity Chamber 47 cu. ft.
SDP
Catalog No. 501893885
Encompass_Preferred
The HH055-SS is a 50 cubic ft double door heated/humidified incubator. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3885 Supplier Darwin Chambers Company Supplier No. HH055SS
Add to Cart
Add to Cart
Request a Quote
The HH055-SS is a 50 cubic ft double door heated/humidified incubator. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.