Learn More
Darwin Chambers Company Triple Door Heat Humidity Chamber 66 cu. ft.
SDP
Supplier:  Darwin Chambers Company HH084SS
Encompass_Preferred
The HH084-SS is a 76.2 cubic ft triple door heated/humidified incubator. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3887
Only null left
Add to Cart
Request a Quote