Learn More
Med Vet International 3M Attest Biological Indicator, Incubator, 116, 1/Case
Catalog No. 502830958
Encompass_Preferred
3M's Attest biological incubator is self-contained to minimize false positives and reduce the risk of contamination 14-vial capacity that is ideal for most office sizes The Attest biological indicator Incubator automatically crushes vials for safety and convenience Color-coded for simple use Dry-block design is preset to 50° C for more consistent and accurate results The translucent cover allows you to monitor the color change indicator
Catalog No. 50-283-0958 Supplier Med Vet International Supplier No. 200071CS
3M's Attest biological incubator is self-contained to minimize false positives and reduce the risk of contamination 14-vial capacity that is ideal for most office sizes The Attest biological indicator Incubator automatically crushes vials for safety and convenience Color-coded for simple use Dry-block design is preset to 50° C for more consistent and accurate results The translucent cover allows you to monitor the color change indicator