Learn More
Chemglass Life Sciences Incubator, 30.8 Cu. Ft., Large Capacity, General Purpose, Reach-In, 120V
SDP
Catalog No. 50143545
Encompass
Horizontal Air FlowFour Interior Electrical OutletsPre-wired Panel for Chart RecorderTempered Glass Viewing Window24 Month Limited Warranty. The CLS-2441 series incubators provide extra large chamber capacities while minimizing the amount of floor space used. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. A totally independent secondary temperature controller offers the added security of over temperature protection. The chamber floors are ruggedly reinforced to support roller apparatus or shakers. Both models are supplied with (6) reinforced, sturdy shelves to support extra heavy loads. Access ports are available by special order. Applications: Hematological Studies Microbiological Determinations Pharmaceutical Stability Assays General Purpose Incubator Applications Large Scale Roller Apparatus Applications Bacterial Culturing and Research Food Processing Quality Control Biochemical Studies
Catalog No. 50-143-545 Supplier Chemglass Life Sciences Supplier No. CLS2441028
Add to Cart
Add to Cart
Request a Quote
Horizontal Air FlowFour Interior Electrical OutletsPre-wired Panel for Chart RecorderTempered Glass Viewing Window24 Month Limited Warranty. The CLS-2441 series incubators provide extra large chamber capacities while minimizing the amount of floor space used. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. A totally independent secondary temperature controller offers the added security of over temperature protection. The chamber floors are ruggedly reinforced to support roller apparatus or shakers. Both models are supplied with (6) reinforced, sturdy shelves to support extra heavy loads. Access ports are available by special order. Applications: Hematological Studies Microbiological Determinations Pharmaceutical Stability Assays General Purpose Incubator Applications Large Scale Roller Apparatus Applications Bacterial Culturing and Research Food Processing Quality Control Biochemical Studies