Learn More
Electron Microscopy Sciences EMS MyTemp™ Mini CO2, 115 V
SDP
Catalog No. 501927919
Encompass_Preferred
At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring carbon dioxide gas control. Its small footprint makes the incubator ideal when bench space is being conserved. Despite its small size, the incubator features a relatively large 20L chamber with the capacity for flasks and bottles up to 2 liters. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour.
Both the temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. Two removable, stainless-steel shelves are included. The shelves are perforated to improve airflow within the chamber.
Catalog No. 50-192-7919 Supplier Electron Microscopy Sciences Supplier No. 8702908
Add to Cart
Add to Cart
Request a Quote
At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring carbon dioxide gas control. Its small footprint makes the incubator ideal when bench space is being conserved. Despite its small size, the incubator features a relatively large 20L chamber with the capacity for flasks and bottles up to 2 liters. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour.
Both the temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. Two removable, stainless-steel shelves are included. The shelves are perforated to improve airflow within the chamber.