Learn More
Chemglass Life Sciences Incubator, MyTemp Mini with Heating and Cooling, 120V
SDP
Catalog No. 501311302
Encompass
Digital temperature control, 13 x 14.5" footprint, Accepts bottles and flasks up to 2L, Internal power outlet for installation of small shakers, mixers, etc. With digital temperature control, the MyTemp incubators provide convenient ¿Set and walk away¿ operation, eliminating the need for external thermometers and repetitive ¿fine tuning¿ of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, -HC model only) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Unique design accepts USA, EU, UK, and AU plugs. Despite a modest footprint of 14.5¿ x 13¿, the Benchmark MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liters. In addition, the incubators include two adjustable/removable shelves for increased capacity. Incubators are CE Approved.
Catalog No. 50-131-1302 Supplier Chemglass Life Sciences Supplier No. CLS245702
Digital temperature control, 13 x 14.5" footprint, Accepts bottles and flasks up to 2L, Internal power outlet for installation of small shakers, mixers, etc. With digital temperature control, the MyTemp incubators provide convenient ¿Set and walk away¿ operation, eliminating the need for external thermometers and repetitive ¿fine tuning¿ of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, -HC model only) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Unique design accepts USA, EU, UK, and AU plugs. Despite a modest footprint of 14.5¿ x 13¿, the Benchmark MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liters. In addition, the incubators include two adjustable/removable shelves for increased capacity. Incubators are CE Approved.