Learn More
Electron Microscopy Sciences EMS MyTemp™ Mini Incubator, Heat / Cool, 230 V, -15°C to 60°C temp range
SDP
Catalog No. 501927915
Encompass_Preferred
This personal sized Mini Digital Incubator with a 13 x 14.5'' footprint accepts bottles and flasks up to 2L. With digital temperature control, the MyTemp incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and the repetitive "fine tuning" of an analog control knob. Includes two adjustable shelves and an internal outlet for powering small shakers, rockers and other instrumentation.
Temperature Range for the heat only model is ambient +1 degree C to 60 degree C.
Temperature Range for the heat and cooling model is ambient Ambient -15 degree C to 60 degree C.
Temp range: -15°C to 60°C
Catalog No. 50-192-7915 Supplier Electron Microscopy Sciences Supplier No. 8702904
This personal sized Mini Digital Incubator with a 13 x 14.5'' footprint accepts bottles and flasks up to 2L. With digital temperature control, the MyTemp incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and the repetitive "fine tuning" of an analog control knob. Includes two adjustable shelves and an internal outlet for powering small shakers, rockers and other instrumentation.
Temperature Range for the heat only model is ambient +1 degree C to 60 degree C.
Temperature Range for the heat and cooling model is ambient Ambient -15 degree C to 60 degree C.
Temp range: -15°C to 60°C