Learn More
Cole-Parmer Stuart Shaking Incubator, 51 L capacity, 230 V
Catalog No. 501960443 Shop All Cole-Parmer Products
Encompass_Preferred
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run. The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC. Order models with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help you comply with regulatory agency requirements.
Catalog No. 50-196-0443 Supplier Cole-Parmer Supplier No. 7952004
Add to Cart
Add to Cart
Request a Quote
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run. The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC. Order models with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help you comply with regulatory agency requirements.