Learn More
Chemglass Life Sciences Chemglass Incubator, Orbital Shaking, 23 Cu Ft, CO2 Humidified
SDP
Catalog No. NC3796803
Encompass
The Chemglass 23 cu ft CO2 humidified orbital shaking incubator is designed for culturing suspension cells and other temperature-sensitive biological samples. It provides precise temperature and CO2 control, orbital shaking across three adjustable shelves, and humidification to minimize medium evaporation.
  • Large 650 L (23 cu ft) chamber capacity.
  • 25 mm orbital stroke for consistent mixing.
  • Temperature range ambient +5 to 60 °C.
  • Temperature uniformity ±0.1 °C at 37 °C.
  • Infrared CO2 sensor with 0-20% control range.
  • Humidified chamber to reduce medium evaporation.
  • Speed range 30-150 rpm.
  • Three removable shelves for high-capacity loading.
  • Exterior dimensions 33.5 x 30.7 x 59 in; interior 27.6 x 25.6 x 59 in.
  • Two-year warranty.
Catalog No. NC3796803 Supplier Chemglass Life Sciences Supplier No. CLS245523
Add to Cart
Add to Cart
Request a Quote
The Chemglass 23 cu ft CO2 humidified orbital shaking incubator is designed for culturing suspension cells and other temperature-sensitive biological samples. It provides precise temperature and CO2 control, orbital shaking across three adjustable shelves, and humidification to minimize medium evaporation.
  • Large 650 L (23 cu ft) chamber capacity.
  • 25 mm orbital stroke for consistent mixing.
  • Temperature range ambient +5 to 60 °C.
  • Temperature uniformity ±0.1 °C at 37 °C.
  • Infrared CO2 sensor with 0-20% control range.
  • Humidified chamber to reduce medium evaporation.
  • Speed range 30-150 rpm.
  • Three removable shelves for high-capacity loading.
  • Exterior dimensions 33.5 x 30.7 x 59 in; interior 27.6 x 25.6 x 59 in.
  • Two-year warranty.