Learn More
Darwin Chambers Company Single Door Thermoelectric Incubator, 33.5"" D x 33.5"" W x 80.25""H
SDP
Catalog No. 501893891
Encompass_Preferred
The TE034 is a 32.1 cubic feet extra wide single door laboratory incubator. Temperature range is 18°C to 50°C. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.
Catalog No. 50-189-3891 Supplier Darwin Chambers Company Supplier No. TE034
Add to Cart
Add to Cart
Request a Quote
The TE034 is a 32.1 cubic feet extra wide single door laboratory incubator. Temperature range is 18°C to 50°C. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.