Learn More
Darwin Chambers Company Double Door Thermoelectric Incubator 47 cu. ft.
SDP
Catalog No. 501893892
Encompass_Preferred
The TE055 is a 50 cubic feet double door laboratory incubator. Temperature range is 20°C to 50°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.
Catalog No. 50-189-3892 Supplier Darwin Chambers Company Supplier No. TE055AA
Add to Cart
Add to Cart
Request a Quote
The TE055 is a 50 cubic feet double door laboratory incubator. Temperature range is 20°C to 50°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.