Learn More
Darwin Chambers Company Triple Door Thermoelectric Incubator 66 cu. ft.
SDP
Supplier:  Darwin Chambers Company TE084AA
Encompass_Preferred
The TE084 is a 76.2 cubic feet triple door laboratory incubator. Temperature range is 20°C to 50°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.
Catalog No. 50-189-3894
Add to Cart
Request a Quote