Learn More
Darwin Chambers Company Benchtop Thermoelectric Incubator 9 cu. ft
SDP
Catalog No. 501893888
Encompass_Preferred
The TE011 is a 11.6 cubic feet benchtop laboratory incubator. Temperature range is 12°C to 50°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.
Catalog No. 50-189-3888 Supplier Darwin Chambers Company Supplier No. TE011
Add to Cart
Add to Cart
Request a Quote
The TE011 is a 11.6 cubic feet benchtop laboratory incubator. Temperature range is 12°C to 50°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.