Learn More
Hach Company Incubator, Hach portable, 12 VDC, 30 - 50 °C (± 0.5 °C)
Supplier:  Hach Company 2569900
Encompass
Field-ready unit can be used with a variety of power sources for reliable, accurate incubation. Operating range: 0 - 40 °C. Accuracy: ±0.5 °C @ 37 °C. Power Source: 12 VDC via vehicle cigarette lighter socket connection. Other Power Source options include 12 VDC rechargeable battery (12 hours of operation @ 37 °C @ 20 - 25 °C ambient) and 115/230 VAC Battery Eliminator. Capacity: 6 P/A bottles or 39 MPN tubes or 42 50 mm petri dishes. External Dimenstion: 30.5 x 30.5 x 25.4 cm (12 x 12 x 10"). Temperature Range: 5 - 50 °C ambient.
Catalog No. 50-199-6254