Learn More
Darwin Chambers Company Single Door Incubator 31 cu. ft.
SDP
Supplier:  Darwin Chambers Company DB030SS
Encompass_Preferred
The DB030SS is a 23.9 cubic feet single door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3905
Add to Cart
Request a Quote