Learn More
Darwin Chambers Company Triple Door Incubator 66 cu. ft.
SDP
Catalog No. 501893910
Encompass_Preferred
The DB084-SS is a 76.2 cubic feet triple door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3910 Supplier Darwin Chambers Company Supplier No. DB084SS
Add to Cart
Add to Cart
Request a Quote
The DB084-SS is a 76.2 cubic feet triple door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.