Learn More
Darwin Chambers Company Single Door Incubator 31 cu. ft.
SDP
Catalog No. 501893904
Encompass_Preferred
The DB030 is a 23.9 cubic feet single door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3904 Supplier Darwin Chambers Company Supplier No. DB030AA
Add to Cart
Add to Cart
Request a Quote
The DB030 is a 23.9 cubic feet single door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.