Learn More
Darwin Chambers Company Single Door Incubator 31 cu. ft.
SDP
Catalog No. 501893906
Encompass_Preferred
The DB034 is a 32.1 cubic feet extra wide single door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3906 Supplier Darwin Chambers Company Supplier No. DB034
Add to Cart
Add to Cart
Request a Quote
The DB034 is a 32.1 cubic feet extra wide single door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.