Learn More
Darwin Chambers Company Double Door Incubator 47 cu. ft.
SDP
Supplier:  Darwin Chambers Company DB055AA
Encompass_Preferred
The DB055 is a 50 cubic feet double door laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3907
Add to Cart
Request a Quote