Learn More
Darwin Chambers Company Benchtop Incubator 9 cu. ft
SDP
Supplier:  Darwin Chambers Company DB011
Encompass_Preferred
The DB011 is a 11.6 cubic feet benchtop laboratory incubator. Temperature range is 2°C to 50°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The DB series laboratory incubators include heating and cooling capabilities, which make them well-suited for both microbiological and BOD studies. These units have lower temperature specifications than our thermoelectrically cooled units, have associated greater heat removal capabilities, and are ideal for locations where the ambient conditions cannot be controlled very well.
Catalog No. 50-189-3903
Add to Cart
Request a Quote