Learn More
Caron Products And Services Refrigerated Incubator, 10 cu. ft., Temp, No RH
Catalog No. NC2200749
Encompass_Preferred
10 ft3 (283 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This benchtop unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
This item is not returnable. View return policy
Catalog No. NC2200749 Supplier Caron Products And Services Supplier No. 7001101
Add to Cart
Add to Cart
Request a Quote
10 ft3 (283 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This benchtop unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.