Learn More
Research Products International Corp Incubator Shaker, Refrigerated, 10LR
SDP
Catalog No. 501976801
Encompass
Incu-Shaker 10LR (Refrigerated) for temperatures down to RT -15°C
Incu-Shaker 10LR is a heavy-duty performer, designed for heavy workloads and continuous use. Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber. Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0.25%, even in the corners.
The powerful brushless induction drive is designed for years of trouble-free service, even under continuous use conditions with heavy loads. Noise and vibration are minimized by an advanced, computer designed motor isolation system. Like other Incu-Shaker models, the 10LR is built to last.
Catalog No. 50-197-6801 Supplier Research Products International Corp Supplier No. 248579
Add to Cart
Add to Cart
Request a Quote
Incu-Shaker 10LR (Refrigerated) for temperatures down to RT -15°C
Incu-Shaker 10LR is a heavy-duty performer, designed for heavy workloads and continuous use. Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber. Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0.25%, even in the corners.
The powerful brushless induction drive is designed for years of trouble-free service, even under continuous use conditions with heavy loads. Noise and vibration are minimized by an advanced, computer designed motor isolation system. Like other Incu-Shaker models, the 10LR is built to last.