Learn More
Med Vet International USA Ready Warm Wall Incubator, J0900R4TWI, Each
Catalog No. 5000495817
Encompass_Preferred
The Ready Warm is a multi-use digital warming device for clinics, laboratories, and research facilities. It is available in 3 models: Wall-mounted Incubator – holds 8 pitri dishes in an enclosure for growing or storing bacterial cultures • Enclosed 4 shelves • 15L capacity • Up to 49°C (120°F) CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: Ambient to 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz ADVANTAGES Compact design Ready Warm Wall Incubator Actual Weight: 1 lb.
Catalog No. 50-004-95817 Supplier Med Vet International Supplier No. JOR900R4TWI
The Ready Warm is a multi-use digital warming device for clinics, laboratories, and research facilities. It is available in 3 models: Wall-mounted Incubator – holds 8 pitri dishes in an enclosure for growing or storing bacterial cultures • Enclosed 4 shelves • 15L capacity • Up to 49°C (120°F) CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: Ambient to 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz ADVANTAGES Compact design Ready Warm Wall Incubator Actual Weight: 1 lb.