Learn More
Yamato Scientific America, Inc. Yamato 567L Incubator with window, General Purpose, 220V
Catalog No. 501898089 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Economical general purpose 567L natural convection incubator with observation window. Dual door system permits contents to be viewed easily without disrupting atmosphere of the incubator. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C
Catalog No. 50-189-8089 Supplier Yamato Scientific America, Inc. Supplier No. IC913CW
Add to Cart
Add to Cart
Request a Quote
Economical general purpose 567L natural convection incubator with observation window. Dual door system permits contents to be viewed easily without disrupting atmosphere of the incubator. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C