Learn More
Yamato Scientific America, Inc. Yamato 97L Incubator with window, General Purpose, 220V
Supplier:  Yamato Scientific America, Inc. IC413CW
Encompass_Preferred
Economical general purpose 97L natural convection incubator with observation window. Dual door system permits contents to be viewed easily without disrupting atmosphere of the incubator. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C
Catalog No. 50-189-8083