Learn More
Thomas Scientific Digital Dry Bath, Single Chamber, without blocks, 230V
SDP
Catalog No. 501460105
Encompass
Digital temperature control (set and walk away) Hinged lid (improves uniformity & conserves energy) External (optional) temperature probe (measures actual sample temperature Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates) Blocks sold separately labForce’s IncuBlock™ Plus series is a “next generation” product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature. The IncuBlock’s transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. It also helps to reduce energy consumption b
Catalog No. 50-146-0105 Supplier Thomas Scientific Supplier No. 1154J45
Digital temperature control (set and walk away) Hinged lid (improves uniformity & conserves energy) External (optional) temperature probe (measures actual sample temperature Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates) Blocks sold separately labForce’s IncuBlock™ Plus series is a “next generation” product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature. The IncuBlock’s transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. It also helps to reduce energy consumption b