Learn More
Chemglass Life Sciences IncuView Integrated Microscope Shelf for 180L SureTherm CO2 Incubator
SDP
Catalog No. 5000083273
Encompass
This integrated microscope shelf is an accessory designed to enable live-cell imaging inside 180 L CO2 incubators. It provides a stable platform for an integrated microscope to observe cell cultures in real time without removing samples from the controlled incubation environment.
  • Supports in situ live-cell imaging within the 180 L incubator chamber.
  • Provides a stable mounting shelf for an integrated microscope.
  • Maintains incubation conditions to minimize contamination during observation.
  • Designed for secure installation and reliable cell viewing.
Catalog No. 50-000-83273 Supplier Chemglass Life Sciences Supplier No. CLS2460185
Add to Cart
Add to Cart
Request a Quote
This integrated microscope shelf is an accessory designed to enable live-cell imaging inside 180 L CO2 incubators. It provides a stable platform for an integrated microscope to observe cell cultures in real time without removing samples from the controlled incubation environment.
  • Supports in situ live-cell imaging within the 180 L incubator chamber.
  • Provides a stable mounting shelf for an integrated microscope.
  • Maintains incubation conditions to minimize contamination during observation.
  • Designed for secure installation and reliable cell viewing.