Learn More
Chemglass Life Sciences IncuView Integrated Microscope Shelf for 45 L SureTherm CO2 Incubator
SDP
Catalog No. 5000083250
Encompass
Optional IncuView integrated microscope shelf for Chemglass 45 L SureTherm CO2 incubators. It enables live-cell imaging inside the incubator so cultures can be observed in real time without removal, helping to minimize temperature fluctuations and lower contamination risk.
  • Enables live-cell imaging without removing cultures from the incubator.
  • Minimizes temperature fluctuations and reduces contamination risk.
  • Designed for compatibility with 45 L reach-in CO2 incubators.
  • Provides a secure, stable mounting platform for integrated microscope components.
  • Removable for maintenance and easy cleaning.
Catalog No. 50-000-83250 Supplier Chemglass Life Sciences Supplier No. CLS2460454
Optional IncuView integrated microscope shelf for Chemglass 45 L SureTherm CO2 incubators. It enables live-cell imaging inside the incubator so cultures can be observed in real time without removal, helping to minimize temperature fluctuations and lower contamination risk.
  • Enables live-cell imaging without removing cultures from the incubator.
  • Minimizes temperature fluctuations and reduces contamination risk.
  • Designed for compatibility with 45 L reach-in CO2 incubators.
  • Provides a secure, stable mounting platform for integrated microscope components.
  • Removable for maintenance and easy cleaning.