Learn More
Cayman Chemical Incyclinide, Purity: ≥98%, CAS: 15866-90-7, 10 mg
SDP
Catalog No. 502948259
Encompass_Preferred
An inhibitor of MMP-13, MMP-1, and MMP-8 (IC50s = 0.3, 34, and 48 µg/ml, respectively); inhibits chick osteoclast digestion of isolated bovine bone at 5 µg/ml and IL-1-induced glycosaminoglycan release from porcine articular cartilage explants at 10 µg/ml; induces cytotoxicity of HeLa and SiHa human cervical cancer cells
Catalog No. 50-294-8259 Supplier Cayman Chemical Supplier No. 3459710 MG
An inhibitor of MMP-13, MMP-1, and MMP-8 (IC50s = 0.3, 34, and 48 µg/ml, respectively); inhibits chick osteoclast digestion of isolated bovine bone at 5 µg/ml and IL-1-induced glycosaminoglycan release from porcine articular cartilage explants at 10 µg/ml; induces cytotoxicity of HeLa and SiHa human cervical cancer cells