Learn More
Medchemexpress LLC HY-13648 10mg , Incyclinide CAS:15866-90-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364810MG
Encompass_Preferred
HY-13648 10mg Incyclinide CAS: 15866-90-7 Incyclinide (CMT-3, COL-3) is a matrix metalloproteinase (MMP) inhibitor, thereby inducing extracellular matrix degradation, and inhibiting angiogenesis, tumor growth and invasion, and metastasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9712