Learn More
MSC CRC T-Force Degreaser, Liquid, Manufacturing Use Only, 5 Gal
Catalog No. 502869244
Encompass
This degreaser quickly dissolves grease, oil, and sludge for efficient operation of mechanical equipment. It evaporates rapidly, leaves no residue, and has a good health and safety profile.
  • Revolutionary formula, free from TCE, Perc, or nPB
  • NSF K1 registered for use in meat and poultry plants
  • Manufacturing use only
  • RoHS compliant
  • Dielectric strength: 29010 V
  • VOC content (federal): 1071 g/L
Catalog No. 50-286-9244 Supplier MSC Supplier No. 53900155
This degreaser quickly dissolves grease, oil, and sludge for efficient operation of mechanical equipment. It evaporates rapidly, leaves no residue, and has a good health and safety profile.
  • Revolutionary formula, free from TCE, Perc, or nPB
  • NSF K1 registered for use in meat and poultry plants
  • Manufacturing use only
  • RoHS compliant
  • Dielectric strength: 29010 V
  • VOC content (federal): 1071 g/L