Learn More
Biosurface Technologies Corp Industrial Surfaces Biofilm Reactor with 220V Stir/Hot Plate, Low- to Mid-Shear Laboratory Biofilm Growth Device, BioSurface Technologies Corporation.
Catalog No. NC3662429
Encompass
"The Industrial Surfaces Biofilm Reactor (ISBR) is a bench-top biofilm growth chamber designed to provide a growth environment for biofilm under low shear, high gas transfer, and intermittent wetting. The sample slides are intermittently wetted via a fluid distribution port in the reactor. The nutrient feed gravity flows into a sump located at the bottom of the reactor and may be recycled or drained to waste. This reactor environment is similar to those in industrial cooling towers, food preparation surfaces, painted and coated surface testing, marine environments, and other periodically wetted or humid biofilm growth surfaces. The ISBR can be used to model biofilm growth found in a large variety of industrial applications and public health environments.The components of the ISBR include: multi-ported glass vessel, ported lid, lid seal assembly, coupon holder rotor, fluid distribution port, 18 coupons (various materials available).
Catalog No. NC3662429 Supplier Biosurface Technologies Corp Supplier No. ISBR 600INT
"The Industrial Surfaces Biofilm Reactor (ISBR) is a bench-top biofilm growth chamber designed to provide a growth environment for biofilm under low shear, high gas transfer, and intermittent wetting. The sample slides are intermittently wetted via a fluid distribution port in the reactor. The nutrient feed gravity flows into a sump located at the bottom of the reactor and may be recycled or drained to waste. This reactor environment is similar to those in industrial cooling towers, food preparation surfaces, painted and coated surface testing, marine environments, and other periodically wetted or humid biofilm growth surfaces. The ISBR can be used to model biofilm growth found in a large variety of industrial applications and public health environments.The components of the ISBR include: multi-ported glass vessel, ported lid, lid seal assembly, coupon holder rotor, fluid distribution port, 18 coupons (various materials available).