Learn More
Uline Industrial Thermal Transfer Labels | Green | SOLD IN CASE QUANTITIES
Catalog No. NC2108945
Encompass
Color code products and equipment for easy tracking and identification, Perforated rolls have smooth surface for excellent printing, Waxresin ribbons recommended for use with color and fluorescent labels, Matte finish paper stock, Adhesive Permanent emulsion acrylic, Perforation Between each label Parallel to the width, Extra wrap of labels added to each roll to compensate for those lost from tape or glue
This item is not returnable. View return policy
Catalog No. NC2108945 Supplier Uline Supplier No. S5956G
Color code products and equipment for easy tracking and identification, Perforated rolls have smooth surface for excellent printing, Waxresin ribbons recommended for use with color and fluorescent labels, Matte finish paper stock, Adhesive Permanent emulsion acrylic, Perforation Between each label Parallel to the width, Extra wrap of labels added to each roll to compensate for those lost from tape or glue