Learn More
Uline Industrial Thermal Transfer Labels - 2 1?2 x 1"", Ribbons Required;
Catalog No. NC2119552
Encompass_Preferred
Industrial Thermal Transfer Labels - 2 1?2 x 1", Ribbons Required; Use with Zebra, Eltron, Fargo, Sato, NEC, Datamax and other thermal transfer printers. Perforated rolls, wound out on a 3" core; 8" outer diameter. Matte finish paper stock; Adhesive: Permanent emulsion acrylic
This item is not returnable. View return policy
Catalog No. NC2119552 Supplier Uline Supplier No. S11292CS
Industrial Thermal Transfer Labels - 2 1?2 x 1", Ribbons Required; Use with Zebra, Eltron, Fargo, Sato, NEC, Datamax and other thermal transfer printers. Perforated rolls, wound out on a 3" core; 8" outer diameter. Matte finish paper stock; Adhesive: Permanent emulsion acrylic