Learn More
MSC Delfin Industrial Vacuum Cleaner, Compressed Air, 45 L, 1 ea
Catalog No. 502819597
Encompass
The Delfin 452 AIR 7V is an industrial vacuum cleaner designed for the suction of dust, solids, and liquids, utilizing compressed air instead of electrical power.
  • Powerful, maintenance-free Venturi unit with 7 or 14 nozzles
  • Large surface polyester filter with manual filter shaker
  • Detachable 45-liter container, with option to use bags
  • Antistatic polyester class M filter with integrated cleaning system
  • Constructed entirely from painted steel
  • Exceptional ratio between air consumption and performance
  • Compact and mobile for versatile applications
Catalog No. 50-281-9597 Supplier MSC Supplier No. 13676374
Add to Cart
Add to Cart
Request a Quote
The Delfin 452 AIR 7V is an industrial vacuum cleaner designed for the suction of dust, solids, and liquids, utilizing compressed air instead of electrical power.
  • Powerful, maintenance-free Venturi unit with 7 or 14 nozzles
  • Large surface polyester filter with manual filter shaker
  • Detachable 45-liter container, with option to use bags
  • Antistatic polyester class M filter with integrated cleaning system
  • Constructed entirely from painted steel
  • Exceptional ratio between air consumption and performance
  • Compact and mobile for versatile applications