Learn More
Cayman Chemical Indacaterol, Purity: ≥98%, CAS: 312753-06-3, 50 mg
SDP
Catalog No. 502981462
Encompass_Preferred
A long-acting β2-adrenergic receptor agonist (EC50 = 8.71 nM for increasing intracellular cAMP levels in vitro); selective for β2-ARs over β1- and β3-ARs (Kis = 43.6, 616.6, and 3,311.3 nM, respectively); inhibits 5-HT-induced bronchoconstriction in conscious guinea pigs with a maximal effect of 85% at 6.7 µg/kg intratracheally; inhibits methacholine-induced bronchoconstriction in rhesus monkeys with a maximal effect of 75% at 12.5 mg/kg via nebulization
Catalog No. 50-298-1462 Supplier Cayman Chemical Supplier No. 2007050 MG
A long-acting β2-adrenergic receptor agonist (EC50 = 8.71 nM for increasing intracellular cAMP levels in vitro); selective for β2-ARs over β1- and β3-ARs (Kis = 43.6, 616.6, and 3,311.3 nM, respectively); inhibits 5-HT-induced bronchoconstriction in conscious guinea pigs with a maximal effect of 85% at 6.7 µg/kg intratracheally; inhibits methacholine-induced bronchoconstriction in rhesus monkeys with a maximal effect of 75% at 12.5 mg/kg via nebulization