Learn More
Cayman Chemical Indacaterol-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2699828-16-3, 1 mg
SDP
Catalog No. 502937557
Encompass_Preferred
An internal standard for the quantification of indacaterol by GC- or LC-MS
Catalog No. 50-293-7557 Supplier Cayman Chemical Supplier No. 288861 MG
An internal standard for the quantification of indacaterol by GC- or LC-MS