Learn More
Selleck Chemical LLC Indacaterol Maleate S3083-1g
SDP
Catalog No. 503109373
Encompass_Preferred
Indacaterol (QAB149) is an ultra-long-acting -adrenoceptor agonist with pKi of 7 36 for 1-adrenoceptor and pKi of 5 48 for 2-adrenoceptor
Catalog No. 50-310-9373 Supplier Selleck Chemical LLC Supplier No. S30831G
Indacaterol (QAB149) is an ultra-long-acting -adrenoceptor agonist with pKi of 7 36 for 1-adrenoceptor and pKi of 5 48 for 2-adrenoceptor