Learn More
Apexbio Technology LLC Indacaterol Maleate 753498-25-8 50mg
SDP
Catalog No. 5000091961
Encompass_Preferred
Indacaterol Maleate (CAS 753498-25-8) is a synthetic ultra-long-acting agonist of -adrenoceptors exhibiting a pK sub i /sub value of 7 36 for these targets By selectively activating sub 2 /sub -adrenergic receptors it modulates intracellular signaling pathways that induce bronchodilation and smooth muscle relaxation Indacaterol Maleate is widely utilized in biomedical research to investigate -adrenergic signaling mechanisms pulmonary pharmacology and novel therapeutic interventions for respiratory disorders involving airway constriction
Catalog No. 50-000-91961 Supplier Apexbio Technology LLC Supplier No. B136950
Indacaterol Maleate (CAS 753498-25-8) is a synthetic ultra-long-acting agonist of -adrenoceptors exhibiting a pK sub i /sub value of 7 36 for these targets By selectively activating sub 2 /sub -adrenergic receptors it modulates intracellular signaling pathways that induce bronchodilation and smooth muscle relaxation Indacaterol Maleate is widely utilized in biomedical research to investigate -adrenergic signaling mechanisms pulmonary pharmacology and novel therapeutic interventions for respiratory disorders involving airway constriction