Learn More
Medchemexpress LLC Indacaterol maleate | 753498-25-8 | 99.6% | C28H32N2O7 | 25 MG
SDP
Catalog No. 5000356971
Encompass_Preferred
Indacaterol maleate (QAB149) is an orally active ultra-long-acting β2 adrenergic receptor (ADRB2) agonist. It inhibits NF-κB activity in a β-arrestin2-dependent manner, preventing lung damage and improving lung function in chronic obstructive pulmonary disorder (COPD). It also shows promise in cardiovascular disease research, normalizing and reversing cardiac remodeling in a myocardial infarction rat model of heart failure.
  • Inhibits NF-κB activity in a β-arrestin2-dependent manner.
  • Helps prevent lung damage.
  • Improves lung function in chronic obstructive pulmonary disorder (COPD).
  • Used in cardiovascular disease research.
  • Normalizes and reverses cardiac remodeling in myocardial infarction rat models.
Catalog No. 50-003-56971 Supplier Medchemexpress LLC Supplier No. 25MGHY14299AR
Indacaterol maleate (QAB149) is an orally active ultra-long-acting β2 adrenergic receptor (ADRB2) agonist. It inhibits NF-κB activity in a β-arrestin2-dependent manner, preventing lung damage and improving lung function in chronic obstructive pulmonary disorder (COPD). It also shows promise in cardiovascular disease research, normalizing and reversing cardiac remodeling in a myocardial infarction rat model of heart failure.
  • Inhibits NF-κB activity in a β-arrestin2-dependent manner.
  • Helps prevent lung damage.
  • Improves lung function in chronic obstructive pulmonary disorder (COPD).
  • Used in cardiovascular disease research.
  • Normalizes and reverses cardiac remodeling in myocardial infarction rat models.