Learn More
Medchemexpress LLC Indacaterol maleate | 753498-25-8 | 99.6% | C28H32N2O7 | 50 MG
SDP
Catalog No. 5000355517
Encompass_Preferred
Indacaterol maleate (Standard) is the analytical standard of Indacaterol maleate and is intended for research and analytical applications. It is an orally active ultra-long-acting β2 adrenergic receptor (ADRB2) agonist that inhibits NF-κB activity in a β-arrestin2-dependent manner. This helps in preventing further lung damage and improving lung function in COPD.
  • Can be used in cardiovascular disease research.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Helps in preventing further lung damage and improving lung function in COPD.
Catalog No. 50-003-55517 Supplier Medchemexpress LLC Supplier No. HY14299AR50MG
Indacaterol maleate (Standard) is the analytical standard of Indacaterol maleate and is intended for research and analytical applications. It is an orally active ultra-long-acting β2 adrenergic receptor (ADRB2) agonist that inhibits NF-κB activity in a β-arrestin2-dependent manner. This helps in preventing further lung damage and improving lung function in COPD.
  • Can be used in cardiovascular disease research.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Helps in preventing further lung damage and improving lung function in COPD.